Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2S | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018063069.1 | XP_017918558.1 | 3 - 4 | 866 | 92 | |
GenBank mRNA | JT703412.1 | - | 4 - 5 | 683 | 92 | |
KA306606.1 | - | 3 - 4 | 731 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |