Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018055744.1 | XP_017911233.1 | 12 - 13 | 3646 | 51 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |