Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAPEPLD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005679103.3 | XP_005679160.1 | 4 - 5 | 1233 | 81 | |
XM_013963249.2 | XP_013818703.2 | 4 - 5 | 1699 | 81 | ||
GenBank mRNA | JO499546.2 | - | 4 - 5 | 1107 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |