Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018056875.1 | XP_017912364.1 | 3 - 4 | 955 | 53 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |