Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005674887.2 | XP_005674944.1 | 1 | 1120 | 87 | |
XM_013976815.1 | XP_013832269.1 | 2 | 1015 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Capra hircus Both primers and probe map within a single exon |