Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CINP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005695419.3 | XP_005695476.2 | 1 - 2 | 61 | 59 | |
XM_018066022.1 | XP_017921511.1 | 1 - 2 | 61 | 59 | ||
XM_018066023.1 | XP_017921512.1 | 1 - 2 | 61 | 59 | ||
GenBank mRNA | JO402024.2 | - | 4 - 5 | 896 | 59 | |
JO443182.2 | - | 1 - 2 | 66 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |