Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013972066.2 | XP_013827520.2 | 7 - 8 | 1564 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |