Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNAI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018061850.1 | XP_017917339.1 | 2 | 204 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Capra hircus Amplicon spans exons and probe does not span exons |