Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYADML2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018065440.1 | XP_017920929.1 | 1 | 1639 | 125 | |
GenBank mRNA | JO555717.2 | - | 1 | 257 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Capra hircus Both primers and probe map within a single exon |