Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agtr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173046.1 | NP_001166517.1 | 1 | 948 | 93 | |
XM_013159551.2 | XP_013015005.1 | 3 | 1336 | 93 | ||
XM_013159553.2 | XP_013015007.1 | 2 | 1103 | 93 | ||
GenBank mRNA | AF165888.1 | - | 1 | 952 | 93 | |
AJ301623.1 | - | 1 | 948 | 93 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Cavia porcellus Both primers and probe map within a single exon |