Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ssp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299140.1 | NP_001286069.1 | 5 - 6 | 4771 | 67 | |
NM_136073.3 | NP_609917.2 | 5 - 6 | 4777 | 67 | ||
GenBank mRNA | BT021951.1 | - | 5 - 6 | 4761 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |