Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnp4F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103413.3 | NP_001096883.3 | 3 - 4 | 788 | 72 | |
NM_001103414.3 | NP_001096884.1 | 4 - 5 | 818 | 72 | ||
NM_078492.4 | NP_511047.2 | 3 - 4 | 995 | 72 | ||
NM_206630.3 | NP_996353.1 | 4 - 5 | 914 | 72 | ||
GenBank mRNA | AY061395.1 | - | 3 - 4 | 1009 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |