Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272322.1 | NP_001259251.1 | 5 - 6 | 2022 | 74 | |
NM_001272323.2 | NP_001259252.1 | 4 - 5 | 1920 | 74 | ||
NM_001272324.2 | NP_001259253.1 | 4 - 5 | 1920 | 74 | ||
NM_001297958.1 | NP_001284887.1 | 4 - 5 | 1920 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |