Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rim2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258911.2 | NP_001245840.1 | 3 - 4 | 792 | 63 | |
NM_001258912.2 | NP_001245841.1 | 3 - 4 | 748 | 63 | ||
NM_134771.4 | NP_608615.1 | 3 - 4 | 813 | 63 | ||
GenBank mRNA | AY058735.1 | - | 3 - 4 | 759 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |