Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tasp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274991.1 | NP_001261920.1 | 2 - 3 | 1096 | 54 | |
NM_140575.4 | NP_648832.1 | 2 - 3 | 1096 | 54 | ||
GenBank mRNA | AY061074.1 | - | 2 - 3 | 1081 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |