Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mi-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014591.3 | NP_001014591.1 | 1 - 2 | 173 | 59 | |
NM_001170005.2 | NP_001163476.1 | 1 - 2 | 173 | 59 | ||
NM_140897.4 | NP_649154.2 | 1 - 2 | 173 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |