Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: osp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144354.3 | NP_001137826.1 | 3 - 4 | 987 | 81 | |
NM_001169504.2 | NP_001162975.1 | 3 - 4 | 987 | 81 | ||
NM_078843.6 | NP_523567.5 | 2 - 3 | 960 | 81 | ||
NM_165089.4 | NP_723879.3 | 2 - 3 | 960 | 81 | ||
GenBank mRNA | BT031256.1 | - | 2 - 3 | 930 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |