Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cact | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273567.1 | NP_001260496.1 | 3 - 4 | 1082 | 76 | |
NM_057594.4 | NP_476942.1 | 3 - 4 | 1027 | 76 | ||
NM_057595.4 | NP_476943.1 | 3 - 4 | 1027 | 76 | ||
NM_165152.2 | NP_723960.1 | 3 - 4 | 976 | 76 | ||
NM_205999.2 | NP_995721.1 | 2 - 3 | 1261 | 76 | ||
GenBank mRNA | AY069399.1 | - | 2 - 3 | 1284 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |