Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dyrk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038817.1 | NP_001033906.1 | 10 - 11 | 1745 | 67 | |
NM_001038818.1 | NP_001033907.1 | 10 - 11 | 1918 | 67 | ||
NM_001273526.1 | NP_001260455.1 | 10 - 11 | 1991 | 67 | ||
NM_001273527.1 | NP_001260456.1 | 6 - 7 | 1338 | 67 | ||
NM_078840.5 | NP_523564.1 | 9 - 10 | 1913 | 67 | ||
NM_205988.3 | NP_995710.1 | 10 - 11 | 2009 | 67 | ||
NM_205989.3 | NP_995711.1 | 10 - 11 | 2005 | 67 | ||
GenBank mRNA | AF168467.1 | - | 10 - 11 | 1980 | 67 | |
BT010088.1 | - | 9 - 10 | 1913 | 67 | ||
BT133412.1 | - | 6 - 7 | 1284 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster Inventoried |