Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: baf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273291.1 | NP_001260220.1 | 1 - 2 | 276 | 82 | |
NM_135332.4 | NP_609176.1 | 1 - 2 | 276 | 82 | ||
GenBank mRNA | BT011552.1 | - | 1 - 2 | 198 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |