Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: qua | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259137.2 | NP_001246066.1 | 5 - 6 | 2851 | 59 | |
NM_165222.2 | NP_724060.1 | 5 - 6 | 2799 | 59 | ||
NM_165223.3 | NP_724061.1 | 5 - 6 | 2839 | 59 | ||
GenBank mRNA | AY121684.1 | - | 5 - 6 | 2790 | 59 | |
BT100209.1 | - | 5 - 6 | 2655 | 59 | ||
U10070.1 | - | 5 - 6 | 2794 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |