Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ninaD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201907.2 | NP_001188836.1 | 1 - 2 | 172 | 88 | |
NM_165237.2 | NP_724087.1 | 1 - 2 | 172 | 88 | ||
GenBank mRNA | BT031187.1 | - | 1 - 2 | 170 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |