Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phlpp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273629.1 | NP_001260558.1 | 2 - 3 | 879 | 69 | |
NM_136094.2 | NP_609938.1 | 2 - 3 | 694 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |