Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pomp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_136213.4 | NP_610057.1 | 2 - 3 | 400 | 53 | |
GenBank mRNA | AY061060.1 | - | 2 - 3 | 366 | 53 | |
KX531769.1 | - | 1 - 2 | 273 | 53 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |