Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: geminin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273798.1 | NP_001260727.1 | 1 | 876 | 98 | |
NM_136374.4 | NP_610218.1 | 2 | 998 | 98 | ||
GenBank mRNA | AF407275.1 | - | 1 | 825 | 98 | |
AY095203.1 | - | 1 | 874 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |