Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsp42Eq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078919.3 | NP_523643.2 | 1 - 2 | 131 | 62 | |
GenBank mRNA | BT150209.1 | - | 1 - 2 | 128 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |