Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: coil | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014505.2 | NP_001014505.1 | 2 - 3 | 259 | 72 | |
NM_001299255.1 | NP_001286184.1 | 2 - 3 | 259 | 72 | ||
GenBank mRNA | BT050478.1 | - | 2 - 3 | 259 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |