Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144173.3 | NP_001137645.1 | 4 - 5 | 970 | 56 | |
NM_001273960.1 | NP_001260889.1 | 2 - 3 | 773 | 56 | ||
NM_136828.3 | NP_610672.1 | 2 - 3 | 773 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |