Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144173.3 | NP_001137645.1 | 7 - 8 | 1394 | 77 | |
NM_001273960.1 | NP_001260889.1 | 5 - 6 | 1176 | 77 | ||
NM_136828.3 | NP_610672.1 | 5 - 6 | 1176 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |