Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Or59c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079099.2 | NP_523823.1 | 1 - 2 | 926 | 86 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Recommended Assay Drosophila melanogaster |