Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Or63a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169860.1 | NP_001163331.1 | 6 - 7 | 1232 | 75 | |
NM_079171.4 | NP_523895.2 | 7 - 8 | 1159 | 75 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Drosophila melanogaster |