Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: norpA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014720.2 | NP_001014720.1 | 14 - 15 | 4196 | 69 | |
NM_001014721.3 | NP_001014721.1 | 13 - 14 | 3734 | 69 | ||
NM_001169190.2 | NP_001162661.1 | 15 - 16 | 4465 | 69 | ||
NM_001297931.1 | NP_001284860.1 | 14 - 15 | 3859 | 69 | ||
NM_080330.4 | NP_525069.2 | 13 - 14 | 3734 | 69 | ||
NM_167008.2 | NP_726925.1 | 14 - 15 | 4196 | 69 | ||
GenBank mRNA | AF181641.1 | - | 13 - 14 | 3717 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |