Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: X11Lbeta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_167239.4 | NP_727440.3 | 10 - 11 | 5887 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |