Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vha16-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_137325.2 | NP_611169.1 | 1 | 469 | 100 | |
GenBank mRNA | BT033048.1 | - | 1 | 469 | 100 | |
KX531398.1 | - | 1 | 423 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |