Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Muc14A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_167482.4 | NP_727928.3 | 430 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |