Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpr65Aw | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_168159.3 | NP_729147.1 | 1 - 2 | 154 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |