Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sclp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169256.1 | NP_001162727.1 | 3 - 4 | 481 | 77 | |
NM_001272546.1 | NP_001259475.1 | 3 - 4 | 813 | 77 | ||
NM_132542.3 | NP_572770.1 | 3 - 4 | 813 | 77 | ||
GenBank mRNA | AY061562.1 | - | 3 - 4 | 813 | 77 | |
BT072930.1 | - | 3 - 4 | 481 | 77 | ||
BT132925.1 | - | 3 - 4 | 813 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |