Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nelf-E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139984.3 | NP_648241.1 | 1 - 2 | 222 | 65 | |
GenBank mRNA | AY071061.1 | - | 1 - 2 | 217 | 65 | |
Y10016.1 | - | 119 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |