Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ND-ACP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274321.1 | NP_001261250.1 | 2 - 3 | 260 | 66 | |
NM_057654.4 | NP_477002.1 | 2 - 3 | 260 | 66 | ||
GenBank mRNA | AY089404.1 | - | 2 - 3 | 403 | 66 | |
KX531823.1 | - | 1 - 2 | 151 | 66 | ||
Y09068.1 | - | 151 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |