Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CalpA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103908.2 | NP_001097378.1 | 14 - 15 | 2545 | 77 | |
NM_001299684.1 | NP_001286613.1 | 15 - 16 | 2590 | 77 | ||
NM_057699.5 | NP_477047.1 | 14 - 15 | 2453 | 77 | ||
GenBank mRNA | AY051678.1 | - | 13 - 14 | 2429 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |