Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dbe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057892.4 | NP_477240.1 | 1 | 1013 | 70 | |
GenBank mRNA | AY069529.1 | - | 1 | 991 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |