Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lcp65Aa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057932.3 | NP_477280.1 | 1 | 78 | 69 | |
GenBank mRNA | BT099966.3 | - | 1 | 49 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |