Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080264.4 | NP_525003.2 | 2 - 3 | 403 | 89 | |
GenBank mRNA | AY071378.1 | - | 2 - 3 | 365 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |