Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mRpL14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272257.2 | NP_001259186.1 | 2 | 531 | 80 | |
NM_080309.4 | NP_525048.1 | 1 | 587 | 80 | ||
GenBank mRNA | AY071662.1 | - | 1 | 565 | 80 | |
AY075539.1 | - | 1 | 575 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |