Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gapdh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038847.2 | NP_001033936.1 | 1 | 1195 | 125 | |
NM_080369.3 | NP_525108.2 | 1 | 1345 | 125 | ||
GenBank mRNA | AY089643.1 | - | 1 | 126 | ||
BT004485.1 | - | 1 | 1220 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Endogenous Controls Recommended Assay Drosophila melanogaster Inventoried |