Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cutlet | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175955.3 | NP_787969.1 | 4 - 5 | 1942 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |