Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irp-1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058023.5 | NP_477371.1 | 5 - 6 | 1171 | 73 | |
GenBank mRNA | AY058657.1 | - | 5 - 6 | 1120 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |