Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mRpL37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079582.5 | NP_524306.3 | 2 - 3 | 607 | 88 | |
GenBank mRNA | BT030992.1 | - | 2 - 3 | 587 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |