Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lbl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275876.2 | NP_001262805.1 | 2 - 3 | 1050 | 78 | |
NM_079710.4 | NP_524434.2 | 2 - 3 | 1050 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |