Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP-1sigma | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260344.2 | NP_001247273.1 | 3 - 4 | 434 | 96 | |
NM_001260345.2 | NP_001247274.1 | 3 - 4 | 434 | 96 | ||
NM_001275963.1 | NP_001262892.1 | 3 - 4 | 434 | 96 | ||
NM_142941.3 | NP_651198.1 | 3 - 4 | 434 | 96 | ||
GenBank mRNA | AY061172.1 | - | 3 - 4 | 382 | 96 | |
BT056295.1 | - | 3 - 4 | 374 | 96 | ||
KX531465.1 | - | 2 - 3 | 289 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |